3-Nitropyrazolo[1,5-a]pyrimidin-5(4H)-one 98%

Supplier: Ambeed
Warning

A802460-100MG A802460-1G A802460-250MG
76653-472EA 27.59 USD
76653-472 76653-474 76653-476
3-Nitropyrazolo[1,5-a]pyrimidin-5(4H)-one 98%
3-Nitropyrazolo[1,5-a]pyrimidin-5(4H)-one

Precaución: for Research Use Only

Formula: C₆H₄N₄O₃
MW: 180.1210 g/mol
MDL Number: MFCD31630873
CAS Number: 1919868-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR