19-(tert-Butoxy)-19-oxononadecanoic acid 97%

Supplier: Ambeed
Warning

A802438-100MG A802438-1G A802438-250MG A802438-5G
76653-464EA 8 USD
76653-464 76653-466 76653-468 76653-470
19-(tert-Butoxy)-19-oxononadecanoic acid 97%
19-(tert-Butoxy)-19-oxononadecanoic acid
Formula: C₂₃H₄₄O₄
MDL Number: MFCD30737821
CAS Number: 1643852-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR