AZ-304 97%

Supplier: Ambeed
Warning

Synonyms: AZ304, 3-(2-Cyanopropan-2-yl)-N-(3-((7-methoxyquinazolin-4-yl)amino)-4-methylphenyl)benzamide

A802126-100MG A802126-10MG A802126-1MG A802126-50MG A802126-5MG
76653-446EA 186.72 USD
76653-446 76653-448 76653-450 76653-452 76653-454
AZ-304 97%
AZ-304

Precaución: for Research Use Only

Formula: C₂₇H₂₅N₅O₂
MW: 451.53 g/mol
MDL Number: MFCD31812677
CAS Number: 942507-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR