1-(5-Fluoro-2-hydroxyphenyl)-2-(3-fluorophenyl)ethanone 98%

Supplier: Ambeed
Warning

A801673-1G A801673-100MG A801673-250MG
76653-432EA 36 USD
76653-432 77784-077 77784-078
1-(5-Fluoro-2-hydroxyphenyl)-2-(3-fluorophenyl)ethanone 98%
1-(5-Fluoro-2-hydroxyphenyl)-2-(3-fluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₂O₂
MDL Number: MFCD29924229
CAS Number: 1300582-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR