3-(3,6-Dibromo-9h-carbazol-9-yl)propanoicacid 95%

Supplier: Ambeed
Warning

A801024-100MG A801024-1G A801024-250MG
76653-426EA 41.38 USD
76653-426 76653-428 76653-430
3-(3,6-Dibromo-9h-carbazol-9-yl)propanoicacid 95%
3-(3,6-Dibromo-9h-carbazol-9-yl)propanoicacid

Precaución: for Research Use Only

Formula: C₁₅H₁₁Br₂NO₂
MW: 397.0613 g/mol
MDL Number: MFCD01176631
CAS Number: 301807-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR