N-Ethynyl-N,4-dimethylbenzenesulfonamide 98%

Supplier: Ambeed
Warning

A800670-1G A800670-250MG A800670-5G A800670-25G
76653-414EA 31 USD
76653-414 76653-416 76653-418 77569-498
N-Ethynyl-N,4-dimethylbenzenesulfonamide 98%
N-Ethynyl-N,4-dimethylbenzenesulfonamide

Precaución: For research use only.

Formula: C₁₀H₁₁NO₂S
MW: 209.2648 g/mol
MDL Number: MFCD30861485
CAS Number: 1005500-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR