N1-Benzyl-N2-(5-methyl-[1,1'-biphenyl]-2-yl)oxalamide 98%

Supplier: Ambeed
Danger

A800664-100MG A800664-1G A800664-250MG A800664-5G
76653-406EA 20.16 USD
76653-406 76653-408 76653-410 76653-412
N1-Benzyl-N2-(5-methyl-[1,1'-biphenyl]-2-yl)oxalamide 98%
N1-Benzyl-N2-(5-methyl-[1,1'-biphenyl]-2-yl)oxalamide

Precaución: for Research Use Only

Formula: C₂₂H₂₀N₂O₂
MW: 344.4064 g/mol
MDL Number: MFCD30491291
CAS Number: 1909333-82-9
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR