tert-Butyl 4-fluoro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%

Supplier: Ambeed
Warning

A799487-100MG A799487-250MG
76653-392EA 61 USD
76653-392 76653-394
tert-Butyl 4-fluoro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate 95%
tert-Butyl 4-fluoro-2-oxospiro[indoline-3,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₁FN₂O₃
MDL Number: MFCD30145814
CAS Number: 2197055-58-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR