4-(DIphenylamino)phenol 95%

Supplier: Ambeed
Warning

A799421-100MG A799421-1G A799421-250MG
76653-386EA 83.82 USD
76653-386 76653-388 76653-390
4-(DIphenylamino)phenol 95%
4-(DIphenylamino)phenol

Precaución: for Research Use Only

Formula: C₁₈H₁₅NO
MW: 261.3178 g/mol
MDL Number: MFCD20259251
CAS Number: 25069-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR