2,2',2'',2'''-((((4-Chlorophenyl)methylene)bis(4,1-phenylene))bis(azanetriyl))tetrakis(ethan-1-ol) 99%

Supplier: Ambeed
Warning

A796121-10MG A796121-1MG A796121-50MG A796121-5MG
76653-352EA 168 USD
76653-352 76653-354 76653-356 76653-358
2,2',2'',2'''-((((4-Chlorophenyl)methylene)bis(4,1-phenylene))bis(azanetriyl))tetrakis(ethan-1-ol) 99%
2,2',2'',2'''-((((4-Chlorophenyl)methylene)bis(4,1-phenylene))bis(azanetriyl))tetrakis(ethan-1-ol)

Precaución: for Research Use Only

Formula: C₂₇H₃₃ClN₂O₄
MDL Number: MFCD00483554
CAS Number: 342777-54-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR