1-(tert-butyl) 6-Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,6-dicarboxylate 98%

Supplier: Ambeed
Warning

A794920-100MG A794920-1G A794920-250MG A794920-25G
76653-328EA 23.34 USD
76653-328 76653-330 76653-332 76653-334
1-(tert-butyl) 6-Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,6-dicarboxylate 98%
1-(tert-butyl) 6-Methyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole-1,6-dicarboxylate

Precaución: for Research Use Only

Formula: C₂₁H₂₈BNO₆
MW: 401.2611 g/mol
MDL Number: MFCD30717679
CAS Number: 2377609-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR