1,3-dimethylimidazolium hexafluorophosphate 99%

Supplier: Ambeed
Warning

A794885-100G A794885-25G A794885-5G
76653-322EA 164.44 USD
76653-322 76653-324 76653-326
1,3-dimethylimidazolium hexafluorophosphate 99%
1,3-dimethylimidazolium hexafluorophosphate

Precaución: for Research Use Only

Formula: C₅H₉F₆N₂P
MW: 242.1025 g/mol
MDL Number: MFCD28015748
CAS Number: 243664-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR