Tin selenide 99.999% (metals basis)

Supplier: Ambeed
Danger

A794614-1G A794614-25G A794614-5G
76653-316EA 7 USD
76653-316 76653-318 76653-320
Tin selenide 99.999% (metals basis)
Tin selenide

Precaución: for Research Use Only

Formula: SeSn
MW: 197.67 g/mol
Melting Pt: 861
MDL Number: MFCD00054049
CAS Number: 1315-06-6
UN: 3283
ADR: 6.1 Ⅲ
Merck Index: 12,08946

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR