MK 212 HCl 98%

Supplier: Ambeed
Danger

Synonyms: MK-212 HCl, 2-Chloro-6-(piperazin-1-yl)pyrazine hydrochloride

A793993-10MG A793993-1MG A793993-25MG A793993-50MG A793993-5MG A793993-250MG
76653-306EA 11.67 USD
76653-306 76653-308 76653-310 76653-312 76653-314 77622-938
MK 212 HCl 98%
MK 212 HCl

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₄
MW: 235.1137 g/mol
MDL Number: MFCD00673963
CAS Number: 61655-58-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR