1-Chloro-2-fluoro-3-isopropoxybenzene 95%

Supplier: Ambeed
Warning

A793923-100MG A793923-1G A793923-250MG
76653-300EA 37.14 USD
76653-300 76653-302 76653-304
1-Chloro-2-fluoro-3-isopropoxybenzene 95%
1-Chloro-2-fluoro-3-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀ClFO
MW: 188.627 g/mol
MDL Number: MFCD23114121
CAS Number: 1369791-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR