3-CHloro-2-methylthiophene 95%

Supplier: Ambeed
Danger

A793496-100MG A793496-1G A793496-250MG
76653-290EA 93.36 USD
76653-290 76653-292 76653-294
3-CHloro-2-methylthiophene 95%
3-CHloro-2-methylthiophene

Precaución: for Research Use Only

Formula: C₅H₅ClS
MW: 132.6112 g/mol
MDL Number: MFCD18415867
CAS Number: 17249-83-1
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR