3-Bromo-N-cyclopropyl-N-(4-methoxybenzyl)benzenesulfonamide 95%

Supplier: Ambeed
Warning

A793453-1G A793453-250MG
76653-286EA 145.35 USD
76653-286 76653-288
3-Bromo-N-cyclopropyl-N-(4-methoxybenzyl)benzenesulfonamide 95%
3-Bromo-N-cyclopropyl-N-(4-methoxybenzyl)benzenesulfonamide

Precaución: for Research Use Only

Formula: C₁₇H₁₈BrNO₃S
MW: 396.2987 g/mol
MDL Number: MFCD10537642
CAS Number: 950163-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR