4-Bromo-N-(3-chloro-2-methylphenyl)benzenesulfonamide 95%

Supplier: Ambeed
Warning

A793405-1G A793405-250MG A793405-5G
76653-280EA 207.94 USD
76653-280 76653-282 76653-284
4-Bromo-N-(3-chloro-2-methylphenyl)benzenesulfonamide 95%
4-Bromo-N-(3-chloro-2-methylphenyl)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrClNO₂S
MW: 360.65 g/mol
MDL Number: MFCD00586864
CAS Number: 311328-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR