(3-Bromo-5-fluorophenyl)(piperidin-1-yl)methanone 95%

Supplier: Ambeed
Warning

A793039-100MG A793039-1G A793039-250MG
76653-260EA 80.63 USD
76653-260 76653-262 76653-264
(3-Bromo-5-fluorophenyl)(piperidin-1-yl)methanone 95%
(3-Bromo-5-fluorophenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₃BrFNO
MW: 286.1401 g/mol
MDL Number: MFCD20541857
CAS Number: 1326982-04-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR