1-Bromo-2,3-dichloro-4-(2-methylpropoxy)benzene 95%

Supplier: Ambeed
Warning

A792904-100MG A792904-1G A792904-250MG
76653-254EA 48 USD
76653-254 76653-256 76653-258
1-Bromo-2,3-dichloro-4-(2-methylpropoxy)benzene 95%
1-Bromo-2,3-dichloro-4-(2-methylpropoxy)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrCl₂O
MDL Number: MFCD29043927
CAS Number: 1881329-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR