3-Chloro-2-(N-ethylaminomethyl)phenylboronic acid, pinacol ester 97%

Supplier: Ambeed
Danger

A792639-1G A792639-250MG A792639-5G
76653-248EA 133.68 USD
76653-248 76653-250 76653-252
3-Chloro-2-(N-ethylaminomethyl)phenylboronic acid, pinacol ester 97%
3-Chloro-2-(N-ethylaminomethyl)phenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₃BClNO₂
MW: 295.6126 g/mol
MDL Number: MFCD20441919
CAS Number: 2096333-72-9
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR