Methyl 2-amino-6-bromonicotinate 95%

Supplier: Ambeed
Warning

A792286-100MG A792286-250MG A792286-1G
76653-234EA 32 USD
76653-234 76653-236 77749-561
Methyl 2-amino-6-bromonicotinate 95%
Methyl 2-amino-6-bromonicotinate

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₂
MDL Number: MFCD19105143
CAS Number: 1227048-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR