1-Bromo-4-chloro-2-propoxybenzene 95%

Supplier: Ambeed
Warning

A792113-100MG A792113-1G A792113-250MG A792113-5G
76653-224EA 37 USD
76653-224 76653-226 76653-228 76653-230
1-Bromo-4-chloro-2-propoxybenzene 95%
1-Bromo-4-chloro-2-propoxybenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
MDL Number: MFCD29043813
CAS Number: 1881331-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR