2,6-Difluoro-3-nitrophenol 95%

Supplier: Ambeed
Danger

A792071-100MG A792071-1G A792071-250MG
76653-218EA 49 USD
76653-218 76653-220 76653-222
2,6-Difluoro-3-nitrophenol 95%
2,6-Difluoro-3-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₃
MDL Number: MFCD28790772
CAS Number: 1393179-72-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR