2-Bromo-4-ethoxybenzaldehyde 95%

Supplier: Ambeed
Warning

A792069-100MG A792069-1G A792069-250MG
76653-212EA 80.63 USD
76653-212 76653-214 76653-216
2-Bromo-4-ethoxybenzaldehyde 95%
2-Bromo-4-ethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD23864715
CAS Number: 861605-18-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR