3-Bromo-2-butoxy-5-chloropyridine 95%

Supplier: Ambeed
Warning

A792039-1G A792039-250MG A792039-5G
76653-206EA 49.87 USD
76653-206 76653-208 76653-210
3-Bromo-2-butoxy-5-chloropyridine 95%
3-Bromo-2-butoxy-5-chloropyridine

Precaución: for Research Use Only

Formula: C₉H₁₁BrClNO
MW: 264.5467 g/mol
MDL Number: MFCD29043799
CAS Number: 1289046-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR