3,4,5-Tris(dodecyloxy)benzaldehyde 98% (by GC)

Supplier: Ambeed
Warning

A791960-100MG A791960-1G A791960-250MG A791960-5G
76653-198EA 15.92 USD
76653-198 76653-200 76653-202 76653-204
3,4,5-Tris(dodecyloxy)benzaldehyde 98% (by GC)
3,4,5-Tris(dodecyloxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₄₃H₇₈O₄
MW: 659.09 g/mol
MDL Number: MFCD29917550
CAS Number: 117241-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR