(3-Bromophenyl)(butyl)sulfane 95%

Supplier: Ambeed
Danger

A791909-100MG A791909-1G A791909-250MG
76653-192EA 44.56 USD
76653-192 76653-194 76653-196
(3-Bromophenyl)(butyl)sulfane 95%
(3-Bromophenyl)(butyl)sulfane

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrS
MW: 245.18 g/mol
MDL Number: MFCD20040716
CAS Number: 156021-46-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR