1-Bromo-4-propylsulfanylbenzene 95%

Supplier: Ambeed
Danger

Synonyms: (4-Bromophenyl)(propyl)sulphane

A791881-1G A791881-250MG A791881-5G
76653-186EA 207.94 USD
76653-186 76653-188 76653-190
1-Bromo-4-propylsulfanylbenzene 95%
1-Bromo-4-propylsulfanylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrS
MW: 231.16 g/mol
MDL Number: MFCD04324355
CAS Number: 76542-19-3
UN: 3335
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR