3-Bromo-6-chloro-2-fluorobenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-Bromo-3-(bromomethyl)-4-chloro-2-fluorobenzene, 6-Chloro-α,3-dibromo-2-fluorotoluene

A791275-100MG A791275-1G A791275-250MG
76653-174EA 53 USD
76653-174 76653-176 76653-178
3-Bromo-6-chloro-2-fluorobenzyl bromide 97%
3-Bromo-6-chloro-2-fluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₄Br₂ClF
MDL Number: MFCD27931488
CAS Number: 886615-32-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR