2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[9H-fluorene] 98% (by HPLC, titration analysis)
Supplier: Ambeed
|
Warning
|
Synonyms:
2,2'-(9,9'-Spirobi[9H-fluorene]-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), (9,9'-Spirobi[9H-fluorene]-2,7-diyl)bis(boronic acid Pinacol ester)
A791208-1G
A791208-250MG
A791208-5G
76653-168EA
45
USD
76653-168
76653-170
76653-172
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[9H-fluorene] 98% (by HPLC, titration analysis)
2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[9H-fluorene]
Precaución: for Research Use Only
Formula:
C₃₇H₃₈B₂O₄ |
CAS Number:
728911-52-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...