N-(5-Aminopyridin-2-yl)-4-(trifluoromethyl)benzamide 97%

Supplier: Ambeed
Warning

Synonyms: TFAP

A790540-10MG A790540-1MG A790540-25MG A790540-5MG
76653-154EA 29 USD
76653-154 76653-156 76653-158 76653-160
N-(5-Aminopyridin-2-yl)-4-(trifluoromethyl)benzamide 97%
N-(5-Aminopyridin-2-yl)-4-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₀F₃N₃O
MW: 281.23 g/mol
MDL Number: MFCD12463359
CAS Number: 1011244-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR