5-Fluoro-4-methyl-2-nitropyridine 95%

Supplier: Ambeed
Warning

A790066-250MG A790066-1G
76653-144EA 172 USD
76653-144 77749-559
5-Fluoro-4-methyl-2-nitropyridine 95%
5-Fluoro-4-methyl-2-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅FN₂O₂
MDL Number: MFCD28798172
CAS Number: 1628184-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR