1,3-Dibromo-2-chloro-5-nitrobenzene 98%

Supplier: Ambeed
Warning

A789766-1G A789766-250MG A789766-5G
76653-138EA 30 USD
76653-138 76653-140 76653-142
1,3-Dibromo-2-chloro-5-nitrobenzene 98%
1,3-Dibromo-2-chloro-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClNO₂
MDL Number: MFCD21605369
CAS Number: 20098-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR