1-(4-(4-(Benzo[d][1,3]dioxole-5-carbonyl)piperazin-1-yl)phenyl)ethanone 98+%

Supplier: Ambeed
Danger

Synonyms: ML406

A789739-10MG A789739-1MG A789739-25MG A789739-50MG A789739-5MG A789739-100MG
76653-128EA 29.71 USD
76653-128 76653-130 76653-132 76653-134 76653-136 77749-556
1-(4-(4-(Benzo[d][1,3]dioxole-5-carbonyl)piperazin-1-yl)phenyl)ethanone 98+%
1-(4-(4-(Benzo[d][1,3]dioxole-5-carbonyl)piperazin-1-yl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₄
MW: 352.38 g/mol
MDL Number: MFCD06004413
CAS Number: 774589-47-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR