(S)-4-(6-Amino-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl)-N-(4-methoxy-2-methylphenyl)-3-methylpiperazine-1-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: UCB9608

A789351-10MG A789351-1MG A789351-50MG A789351-5MG
76653-120EA 66.84 USD
76653-120 76653-122 76653-124 76653-126
(S)-4-(6-Amino-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl)-N-(4-methoxy-2-methylphenyl)-3-methylpiperazine-1-carboxamide 98%
(S)-4-(6-Amino-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl)-N-(4-methoxy-2-methylphenyl)-3-methylpiperazine-1-carboxamide

Precaución: for Research Use Only

Formula: C₂₀H₂₆N₈O₂
MW: 410.47 g/mol
MDL Number: MFCD01744844
CAS Number: 1616413-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR