2,3,4-Trimethylaniline 97%

Supplier: Ambeed
Warning

A787649-100MG A787649-1G A787649-250MG
76653-088EA 49.87 USD
76653-088 76653-090 76653-092
2,3,4-Trimethylaniline 97%
2,3,4-Trimethylaniline

Precaución: for Research Use Only

Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00130021
CAS Number: 1467-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR