4-Aminobicyclo[2.2.2]octane-1-carboxamide 98%

Supplier: Ambeed
Warning

A786165-100MG A786165-250MG A786165-50MG
76653-072EA 235.52 USD
76653-072 76653-074 76653-076
4-Aminobicyclo[2.2.2]octane-1-carboxamide 98%
4-Aminobicyclo[2.2.2]octane-1-carboxamide

Precaución: for Research Use Only

Formula: C₉H₁₆N₂O
MW: 168.2361 g/mol
MDL Number: MFCD20702135
CAS Number: 863303-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR