(S)-1-Benzyl 4-methyl 2-((tert-butoxycarbonyl)amino)succinate 95%

Supplier: Ambeed
Warning

A784665-100MG A784665-1G A784665-250MG
76653-058EA 26 USD
76653-058 76653-060 76653-062
(S)-1-Benzyl 4-methyl 2-((tert-butoxycarbonyl)amino)succinate 95%
(S)-1-Benzyl 4-methyl 2-((tert-butoxycarbonyl)amino)succinate

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₆
MW: 337.3676 g/mol
MDL Number: MFCD27578330
CAS Number: 158201-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR