(2R,3R)-Dimethyl 2,3-bis((4-methylbenzoyl)oxy)succinate 95%

Supplier: Ambeed
Warning

A784077-1G A784077-250MG
76653-054EA 346 USD
76653-054 76653-056
(2R,3R)-Dimethyl 2,3-bis((4-methylbenzoyl)oxy)succinate 95%
(2R,3R)-Dimethyl 2,3-bis((4-methylbenzoyl)oxy)succinate

Precaución: for Research Use Only

Formula: C₂₂H₂₂O₈
MW: 414.4053 g/mol
MDL Number: MFCD27578184
CAS Number: 2703746-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR