Potassium trifluoro[3-(methoxycarbonyl)phenyl]boranuide 98%

Supplier: Ambeed
Warning

A783956-1G A783956-5G
76653-050EA 33 USD
76653-050 76653-052
Potassium trifluoro[3-(methoxycarbonyl)phenyl]boranuide 98%
Potassium trifluoro[3-(methoxycarbonyl)phenyl]boranuide

Precaución: for Research Use Only

Formula: C₈H₇BF₃KO₂
MDL Number: MFCD09993571
CAS Number: 1242733-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR