Donepezil hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Benzyl-4-piperidylmethyl)-5,6-dimethoxy-1-indanone Hydrochloride, 2-((1-Benzylpiperidin-4-yl)methyl)-5,6-dimethoxy-2,3-dihydro-1H-inden-1-one hydrochloride

A783895-1G A783895-25G A783895-5G
76653-044EA 5.31 USD
76653-044 76653-046 76653-048
Donepezil hydrochloride 98%
Donepezil hydrochloride

Precaución: for Research Use Only

Formula: C₂₄H₂₉NO₃
MW: 415.96 g/mol
MDL Number: MFCD00881312
CAS Number: 120011-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR