Nalidixic acid sodium salt 98%

Supplier: Ambeed
Danger

A783659-100MG A783659-1G A783659-1MG A783659-25G A783659-5G
76653-034EA 7 USD
76653-034 76653-036 76653-038 76653-040 76653-042
Nalidixic acid sodium salt 98%
Nalidixic acid sodium salt

Precaución: for Research Use Only

Formula: C₁₂H₁₁N₂NaO₃
MDL Number: MFCD00064376
CAS Number: 3374-05-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR