(1H-Pyrrolo[3,2-c]pyridin-2-yl)methanamine dihydrochloride 95%

Supplier: Ambeed
Warning

A782671-100MG A782671-250MG A782671-1G
76653-022EA 61.54 USD
76653-022 77618-152 77749-546
(1H-Pyrrolo[3,2-c]pyridin-2-yl)methanamine dihydrochloride 95%
(1H-Pyrrolo[3,2-c]pyridin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁Cl₂N₃
MW: 220.0990 g/mol
MDL Number: MFCD30537042
CAS Number: 2408959-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR