SLV-2436 97%

Supplier: Ambeed
Warning

Synonyms: 5-(3-Amino-1H-indazol-6-yl)-1-(3-chlorobenzyl)pyridin-2(1H)-one

A781993-10MG A781993-1MG A781993-5MG
76653-010EA 37.14 USD
76653-010 76653-012 76653-014
SLV-2436 97%
SLV-2436

Precaución: for Research Use Only

Formula: C₁₉H₁₅ClN₄O
MW: 350.8 g/mol
MDL Number: MFCD31728203
CAS Number: 2095704-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR