(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(7-methyl-1H-indol-3-yl)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Trp(7-Me)-OH

A779029-25G A779029-5G A779029-1G A779029-250MG
76652-992EA 1102 USD
76652-992 76652-994 77595-242 77749-536
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(7-methyl-1H-indol-3-yl)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(7-methyl-1H-indol-3-yl)propanoic acid

Precaución: for Research Use Only

Formula: C₂₇H₂₄N₂O₄
MW: 440.4905 g/mol
MDL Number: MFCD31560627
CAS Number: 1808268-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR