1-(Cyclohexylcarbonyl)piperidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A778824-1G A778824-250MG
76652-982EA 30.77 USD
76652-982 76652-984
1-(Cyclohexylcarbonyl)piperidine-3-carboxylic acid 95%
1-(Cyclohexylcarbonyl)piperidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₁NO₃
MW: 239.3107 g/mol
MDL Number: MFCD03128019
CAS Number: 436093-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR