2,3-Difluoro-5-formylphenylboronic acid 96%

Supplier: Ambeed
Warning

A778103-1G A778103-250MG A778103-5G
76652-970EA 74.27 USD
76652-970 76652-972 76652-974
2,3-Difluoro-5-formylphenylboronic acid 96%
2,3-Difluoro-5-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BF₂O₃
MW: 185.92 g/mol
MDL Number: MFCD16251600
CAS Number: 2377611-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR