7-Fluoro-1-isopropyl-3-methyl-8-(6-(3-(piperidin-1-yl)propoxy)pyridin-3-yl)-1,3-dihydro-2H-imidazo[4,5-c]quinolin-2-one 98%

Supplier: Ambeed
Warning

Synonyms: AZD1390

A775616-10MG A775616-1MG A775616-25MG A775616-5MG
76652-948EA 357.53 USD
76652-948 76652-950 76652-952 76652-954
7-Fluoro-1-isopropyl-3-methyl-8-(6-(3-(piperidin-1-yl)propoxy)pyridin-3-yl)-1,3-dihydro-2H-imidazo[4,5-c]quinolin-2-one 98%
7-Fluoro-1-isopropyl-3-methyl-8-(6-(3-(piperidin-1-yl)propoxy)pyridin-3-yl)-1,3-dihydro-2H-imidazo[4,5-c]quinolin-2-one

Precaución: for Research Use Only

Formula: C₂₇H₃₂FN₅O₂
MW: 477.57 g/mol
MDL Number: MFCD31619275
CAS Number: 2089288-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR