1,3-Dimesitylimidazolium-2-carboxylate 98% (by HPLC, titration analysis)
Supplier: Ambeed
|
Warning
|
Synonyms:
IMes.CO2, 1,3-Bis(2,4,6-trimethylphenyl)imidazolium-2-carboxylate
A775276-1G
A775276-250MG
A775276-5G
76652-942EA
84.88
USD
76652-942
76652-944
76652-946
1,3-Dimesitylimidazolium-2-carboxylate 98% (by HPLC, titration analysis)
1,3-Dimesitylimidazolium-2-carboxylate
Precaución: for Research Use Only
Formula:
C₂₂H₂₄N₂O₂ MW: 348.45 g/mol |
MDL Number:
MFCD31618131 CAS Number: 675877-56-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...